Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgM, Clone: G53-238, biotin, Secondary Antibody, BD

Catalog No. BDB553886 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553886 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553886 Supplier BD Biosciences Supplier No. 553886
Add to Cart
Add to Cart

The G53-238 antibody reacts specifically with rat IgM monomers and pentamers. It does not react with other Ig isotypes. G53-238 antibody has not been shown to stimulate B-cell proliferation.

Host Species: Mouse
Clone: G53-238
Isotype: IgG1, κ
Species Reactivity: Rat
Immunogen: Pooled rat Ig

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat IgM
Classification Monoclonal
Clone G53-238
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Mouse
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rat
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.