Learn More
Mouse anti-Rat IgM, Clone: G53-238, FITC, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The G53-238 antibody reacts specifically with rat IgM monomers and pentamers. It does not react with other Ig isotypes. G53-238 antibody has not been shown to stimulate B-cell proliferation.
Host Species: Mouse
Clone: G53-238
Isotype: IgG1, κ
Species Reactivity: Rat
Immunogen: Pooled rat Ig
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Rat IgM |
| Classification | Monoclonal |
| Clone | G53-238 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.