Learn More
Invitrogen™ Mouse anti-Rat IgM (Heavy chain) Secondary Antibody, FITC
Shop All Thermo Scientific ProductsDescription
For FACS analysis, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat IgM Heavy Chain antibody, clone MARM-4 recognizes the mu heavy chain of rat immunoglobulin and does not cross-react with other immunoglobulin classes.
Specifications
Specifications
| Antigen | Rat IgM (Heavy chain) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MARM-4 |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide |
| Host Species | Mouse |
| Immunogen | Polyclonal rat IgM. |
| Purification Method | Affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.