Learn More
Mouse anti-Rat IgM Heavy Chain Secondary Antibody (MARM-4), PE, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgM Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MARM-4 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain |
| Host Species | Mouse |
| Immunogen | IR202 rat IgM myeloma protein |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.