Learn More
Invitrogen™ Mouse anti-Rat Kappa Light Chain Secondary Antibody, FITC
Shop All Thermo Scientific ProductsDescription
Mouse anti-Rat Kappa Light Chain Secondary Antibody for Flow
Specifications
Specifications
| Antigen | Rat Kappa Light Chain |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OX-12 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| Host Species | Mouse |
| Immunogen | Pooled rat serum IgG. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.