Learn More
Biolegend Purified anti-V5-tag
Supplier:  Biolegend 680602
Encompass_Preferred
Purified anti-V5-tag [7/4]; Isotype: Mouse IgG2b, κ; Reactivity: GKPIPNPLLGLDST tag epitope; Apps: WB, IP; Size: 100 ug
Catalog No. NC1040697