Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 9 (MMP9) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11904
Encompass_Preferred
Mouse Matrix Metalloproteinase 9 (MMP9) Anti-antigen ELISA Kit for detecting MMP9 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.13ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P41245. Gene ID: 17395. Target Synonyms: GELB; Gelatinase B; CLG4B; CLG4-B; 92 KDa Gelatinase; 92kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6926