Learn More
Biomatik Corporation Mouse Anti-Mullerian Hormone (AMH) CLIA Kit
Supplier:  Biomatik Corporation EKU11555
Encompass_Preferred
Mouse Anti-Mullerian Hormone (AMH) CLIA Kit for detecting AMH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 39.06-10,000pg/ml. Sensitivity: 14.13pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P27106. Gene ID: 11705. Target Synonyms: MIF; MIH; MIS; Müllerian Inhibiting Factor; Müllerian Inhibiting Hormone; Müllerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7664