Learn More
Biomatik Corporation Mouse Anti-Mullerian Hormone (AMH) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12485
Encompass_Preferred
Mouse Anti-Mullerian Hormone (AMH) Mini sample ELISA Kit for detecting AMH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 48.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27106. Gene ID: 11705. Target Synonyms: MIF; MIH; MIS; Müllerian Inhibiting Factor; Müllerian Inhibiting Hormone; Müllerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7597