Learn More
Biomatik Corporation Mouse Thyroglobulin (TG) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12127
Encompass_Preferred
Mouse Thyroglobulin (TG) Anti-antigen ELISA Kit for detecting TG in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.21ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: O08710. Gene ID: 21819. Target Synonyms: AITD3; TGN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8024