Learn More
BIOHIPPO Mouse APC Microsample
SDP
Catalog No. 5000315304
Encompass
Mouse APC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APC in plasma It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 32 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse APC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse APC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse APC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15304 Supplier BIOHIPPO Supplier No. BHE15211861
Mouse APC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APC in plasma It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 32 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse APC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse APC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse APC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color