Learn More
Sino Biological Anti-Serum Amyloid P Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50113RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Serum amyloid P component (Catalog#50113-M08H; NP_035448.2; Met1-Asp224). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2492