Learn More
Sino Biological Recombinant Mouse Serum Amyloid P Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50113M08H1
Encompass_Preferred
A DNA sequence encoding the mouse APCS (NP_035448.2) (Met 1-Asp 224) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-241-2486