Learn More
Sino Biological Anti-APLP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51730T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse APLP1 / Amyloid-like protein 1 (rM APLP1 / Amyloid-like protein 1; Catalog#51730-M08H; NP_031493.2; Met1-Glu584). APLP1 / Amyloid-like protein 1 specific IgG was purified by Mouse APLP1 / Amyloid-like protein 1 affinity chromatography.
Catalog No. 50-255-2967