Learn More
Sino Biological Anti-Apolipoprotein A-I/APOA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50918T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ApoA1 (Catalog#50918-M08H; Q00623; Met1-Gln264). ApoA1 specific IgG was purified by Mouse ApoA1 affinity chromatography.
Catalog No. 50-241-1938