Learn More
Sino Biological Recombinant Mouse Apolipoprotein A-I/APOA1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 50918M02H1100
Encompass_Preferred
A DNA sequence encoding the Mouse APOA1 (Q00623) (Asp25-Gln264) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-241-1815