Learn More
Bon Opus Biosciences Recombinant Mouse Apolipoprotein E/ApoE (C-His) - Source: Human 293 Cells
SDP
Catalog No. NC2733658
Encompass_Preferred
Recombinant Mouse Apolipoprotein E is produced by our Mammalian expression system and the target gene encoding Glu19-Gln311 is expressed with a 6His tag at the C-terminus. Accession#P08226. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 0.1%Tween 80, 10% trehalose, pH 7.4
Catalog No. NC2733658 Supplier Bon Opus Biosciences Supplier No. BP20550UG
Recombinant Mouse Apolipoprotein E is produced by our Mammalian expression system and the target gene encoding Glu19-Gln311 is expressed with a 6His tag at the C-terminus. Accession#P08226. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 0.1%Tween 80, 10% trehalose, pH 7.4