Learn More
BIOHIPPO Mouse APOH Microsample
SDP
Catalog No. 5000315257
Encompass
Mouse APOH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APOH in Serum plasma saliva and other biological fluids It provides a detection range of 7 82-500 g/mL with a sensitivity of 1 8 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse APOH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse APOH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse APOH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15257 Supplier BIOHIPPO Supplier No. BHE15212704
Mouse APOH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APOH in Serum plasma saliva and other biological fluids It provides a detection range of 7 82-500 g/mL with a sensitivity of 1 8 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse APOH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse APOH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse APOH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color