Learn More
Biomatik Corporation Mouse Apolipoprotein A1 (APOA1) CLIA Kit
Supplier:  Biomatik Corporation EKU09770
Encompass_Preferred
Mouse Apolipoprotein A1 (APOA1) CLIA Kit for detecting APOA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q00623. Gene ID: 11806. Target Synonyms: Apo-A1; ApoA-1 Milano; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8440