Learn More
Biomatik Corporation Mouse Apolipoprotein A5 (APOA5) ELISA Kit
Supplier:  Biomatik Corporation EKU02496
Encompass_Preferred
Mouse Apolipoprotein A5 (APOA5) ELISA Kit for detecting APOA5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.139ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8C7G5. Target Synonyms: Apo-A5; APOA-V; APOAV; RAP3; Regeneration-associated protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5206