Learn More
Biomatik Corporation Mouse Apolipoprotein B (APOB) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10520
Encompass_Preferred
Mouse Apolipoprotein B (APOB) Wide-range ELISA Kit for detecting APOB in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000ng/ml. Sensitivity: 13.5ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: E9Q414. Gene ID: 238055. Target Synonyms: Apo-B; FLDB; Apo B-100; Apo B-48. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5387