Learn More
Biomatik Corporation Mouse Apolipoprotein B (Apob) Elisa Kit
Supplier:  Biomatik Corporation EKC31381
Encompass_Preferred
Mouse Apolipoprotein B (Apob) Elisa Kit for detecting Apolipoprotein B in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ug/ml. Sensitivity: 0.039ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: E9Q414; Apob. Target Synonyms: ApobApolipoprotein B-100; Apo B-100) [Cleaved into: Apolipoprotein B-48; Apo B-48)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6646