Learn More
Biomatik Corporation Mouse Apolipoprotein C1 (APOC1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12405
Encompass_Preferred
Mouse Apolipoprotein C1 (APOC1) Mini sample ELISA Kit for detecting APOC1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34928. Gene ID: 11812. Target Synonyms: Apo-C1; Truncated apolipoprotein C-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7970