Learn More
ACROBiosystems Mouse Apolipoprotein E Protein, His Tag (MALS & SPR verified)
Catalog No. 502331781
Encompass_Preferred
Mouse Apolipoprotein E Protein, His Tag (APE-M52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Gln 311 (Accession # P08226).
Catalog No. 50-233-1781 Supplier ACROBiosystems Supplier No. APEM52H5100UG
Mouse Apolipoprotein E Protein, His Tag (APE-M52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Gln 311 (Accession # P08226).