Learn More
American Research Products Inc Mouse Apoptosis associated spe
Catalog No. 5000328347
Encompass
Mouse Apoptosis-associated speck-like protein containing a CARD (PYCARD) ELISA kit from CUSABIO Detection Range 7 81 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28347 Supplier American Research Products Inc Supplier No. CSBEL019114MO
Mouse Apoptosis-associated speck-like protein containing a CARD (PYCARD) ELISA kit from CUSABIO Detection Range 7 81 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich