Learn More
Biomatik Corporation Mouse Apoptotic Peptidase Activating Factor 1 (APAF1) ELISA Kit
Supplier:  Biomatik Corporation EKU09043
Encompass_Preferred
Mouse Apoptotic Peptidase Activating Factor 1 (APAF1) ELISA Kit for detecting APAF1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88879. Gene ID: 11783. Target Synonyms: CED4; Apoptotic Protease Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8574