Learn More
Biomatik Corporation Mouse Apoptotic Peptidase Activating Factor 1 (APAF1) CLIA Kit
Supplier:  Biomatik Corporation EKU09657
Encompass_Preferred
Mouse Apoptotic Peptidase Activating Factor 1 (APAF1) CLIA Kit for detecting APAF1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.87pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O88879. Gene ID: 11783. Target Synonyms: CED4; Apoptotic Protease Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4624