Learn More
Kingfisher Biotech Inc Mouse APRIL Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0360M005
Encompass
The Mouse APRIL yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse APRIL applications are for cell culture, ELISA standard, and Western Blot Control. The Mouse APRIL yeast-derived recombinant protein can be purchased in multiple sizes. Mouse APRIL Specifications: (Molecular Weight: 16.4 kDa) (Amino Acid Sequence: AVLTQKHKKKHSVLHLVPVNITSKADSDVTEVMWQPVLRRGRGLEAQGDIVRVWDTGIYLLYSQVLFHDVTFTMGQVVSREGQGRRETLFRCIRSMPSDPDRAYNSCYSAGVFHLHQGDIITVKIPRANAKLSLSPHGTFLGFVKL (146)) (Gene ID: 69583). For research use only. Made in the USA
Catalog No. 50-241-3729