Learn More
Biomatik Corporation Mouse Aquaporin 3, Gill Blood Group (AQP3) ELISA Kit
Supplier:  Biomatik Corporation EKU10659
Encompass_Preferred
Mouse Aquaporin 3, Gill Blood Group (AQP3) ELISA Kit for detecting AQP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R2N1. Gene ID: 11828. Target Synonyms: GIL; Aquaglyceroporin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6794