Learn More
Biomatik Corporation Mouse Arachidonate-12-Lipoxygenase (ALOX12) ELISA Kit
Supplier:  Biomatik Corporation EKU12318
Encompass_Preferred
Mouse Arachidonate-12-Lipoxygenase (ALOX12) ELISA Kit for detecting ALOX12 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P39655. Gene ID: 11684. Target Synonyms: LOX12; 12-LOX; ALOX-12; 12-LO; 12LPG; LOG12; Lipoxygenase 12; 12S-lipoxygenase; Lipoxin synthase 12-LO; Platelet-type lipoxygenase 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7985