Learn More
BIOHIPPO Mouse ARC Microsample
SDP
Catalog No. 5000314568
Encompass
Mouse ARC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ARC in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ARC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ARC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ARC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14568 Supplier BIOHIPPO Supplier No. BHE15211146
Mouse ARC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ARC in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ARC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ARC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ARC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color