Learn More
Biomatik Corporation Mouse Arginase (ARG) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11078
Encompass_Preferred
Mouse Arginase (ARG) Mini sample ELISA Kit for detecting ARG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61176. Gene ID: 11846. Target Synonyms: ARG1; Arginase I; Liver Arginase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5948