Learn More
Sino Biological Recombinant Anti-ARSA Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50018R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Arylsulfatase A / ARSA (rM Arylsulfatase A / ARSA; Catalog#50018-M08H; NP_033843.2; Met 1-Ser 506).
Catalog No. 50-255-3552