Learn More
SAI Infusion Technologies Mouse Arterial Catheter - 15cm PU
SDP
Catalog No. NC9561357
Encompass
Arterial Catheter for Mice 25g and over. 15cm PU catheter with 10mm 1F tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 25G blunt needle (such as B25-50). Product is for laboratory animal research only.
Catalog No. NC9561357 Supplier SAI Infusion Technologies Supplier No. MAC2B
Arterial Catheter for Mice 25g and over. 15cm PU catheter with 10mm 1F tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 25G blunt needle (such as B25-50). Product is for laboratory animal research only.