Learn More
Cayman Chemical Mouse Aryl Hydrocarbon Receptor Reporter Assay System, 3x32 Well
SDP
Catalog No. 502915014
Encompass_Preferred
INDIGO’s Mouse AhR assay kit is an all-inclusive system. In addition to Mouse AhR Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist MeBIO, Luciferase Detection Reagent, and a cell culture-ready assay plate.
Catalog No. 50-291-5014 Supplier Cayman Chemical Supplier No. 369043X32 WELL
INDIGO’s Mouse AhR assay kit is an all-inclusive system. In addition to Mouse AhR Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist MeBIO, Luciferase Detection Reagent, and a cell culture-ready assay plate.