Learn More
Biomatik Corporation Mouse Aryl Hydrocarbon Receptor (AhR) ELISA Kit
Supplier:  Biomatik Corporation EKU10664
Encompass_Preferred
Mouse Aryl Hydrocarbon Receptor (AhR) ELISA Kit for detecting AhR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30561. Gene ID: 11622. Target Synonyms: BHLHE76; Class E basic helix-loop-helix protein 76. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4988