Learn More
As One International Inc Mouse Adipose-derived Stem Cell Growth Medium, 1kit
Catalog No. 501655022
Encompass_Preferred
Developed for the optimal proliferation of adipose-derived stem cells, allowing them to expand through multiple passages while retaining the ability to differentiate into osteoblasts, adipocytes and chondrocytes
Catalog No. 50-165-5022 Supplier As One International Inc Supplier No. MUXMD90011
Developed for the optimal proliferation of adipose-derived stem cells, allowing them to expand through multiple passages while retaining the ability to differentiate into osteoblasts, adipocytes and chondrocytes