Learn More
Sino Biological Recombinant Anti-ASGR1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50083R103100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ASGPR1 / ASGR1 (rM ASGPR1 / ASGR1; Catalog#50083-M07H; NP_033844.1; Ser 60-Asn 284).
Catalog No. 50-241-2030