Learn More
Sino Biological Anti-ASGR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50083T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ASGR1 (rM ASGR1; Catalog#50083-M07H; NP_033844.1; Ser60-Asn284). ASGR1 specific IgG was purified by Mouse ASGR1 affinity chromatography.
Catalog No. 50-255-3476