Learn More
Sino Biological Anti-ASGR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50083T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ASGPR1 (rM ASGPR1 ; Catalog#50083-M07H; NP_033844.1; Ser60-Asn284). ASGPR1 specific IgG was purified by Mouse ASGPR1 affinity chromatography.
Catalog No. 50-241-0277