Learn More
Sino Biological Anti-Argininosuccinate lyase Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106760T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse Argininosuccinate lyase, and purified by antigen affinity chromatography.
Catalog No. 50-241-2564