Learn More
BIOHIPPO Mouse AST Microsample
SDP
Catalog No. 5000314527
Encompass
Mouse AST Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of AST in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 U/L with a sensitivity of 1 34 U/L The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse AST Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse AST Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse AST biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14527 Supplier BIOHIPPO Supplier No. BHE15212199
Mouse AST Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of AST in serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 U/L with a sensitivity of 1 34 U/L The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse AST Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse AST Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse AST biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color