Learn More
Sciencell Research Laboratories MOUSE ASTROCYTES
Supplier:  Sciencell Research Laboratories M180057
Encompass
Isolated from postnatal day 2 C57BL/6 mouse brain. MA are cryopreserved as primary cultures and delivered frozen. Each vial contains >5 x 10^5 cells in 1 ml volume.
Catalog No. NC0264905