Learn More
Raybiotech Inc C-Series Mouse Atherosclerosis Antibody Array 1 Kit.
Supplier: Raybiotech Inc AAMATH14
Detects 22 Mouse Atherosclerotic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: bFGF, CD40 (TNFRSF5), Eotaxin-1 (CCL11), GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-3, IL-4, IL-5, IL-6, IL-13, L-Selectin (CD62L), MCP-1 (CCL2), M-CSF, MIP-3 alpha (CCL20), P-Selectin, RANTES (CCL5), TNF alpha, VEGF-A