Learn More
Biomatik Corporation Mouse ATP Binding Cassette Transporter A9 (ABCA9) ELISA Kit
Supplier:  Biomatik Corporation EKU09173
Encompass_Preferred
Mouse ATP Binding Cassette Transporter A9 (ABCA9) ELISA Kit for detecting ABCA9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K449. Gene ID: 217262. Target Synonyms: ABC-A9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6867