Learn More
Biomatik Corporation Mouse ATP Binding Cassette Transporter G2 (ABCG2) ELISA Kit
Supplier:  Biomatik Corporation EKU02623
Encompass_Preferred
Mouse ATP Binding Cassette Transporter G2 (ABCG2) ELISA Kit for detecting ABCG2 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7TMS5. Gene ID: 26357. Target Synonyms: CDw338; CD338; ABC-G2; MRX; BCRP1; ABC15; ABCP; BCRP; BMDP; MXR; MXR1; Mitoxantrone resistance-associated protein; Breast Cancer Resistance Protein 1; Urate exporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7019