Learn More
Biomatik Corporation Mouse ATP Binding Cassette Transporter A13 (ABCA13) ELISA Kit
Supplier:  Biomatik Corporation EKU02606
Encompass_Preferred
Mouse ATP Binding Cassette Transporter A13 (ABCA13) ELISA Kit for detecting ABCA13 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5SSE9. Gene ID: 268379. Target Synonyms: ABC-A13; ATP-binding cassette sub-family A member 13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9113