Learn More
Biomatik Corporation Mouse ATP Binding Cassette Transporter C6 (ABCC6) ELISA Kit
Supplier:  Biomatik Corporation EKU02618
Encompass_Preferred
Mouse ATP Binding Cassette Transporter C6 (ABCC6) ELISA Kit for detecting ABCC6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R1S7. Gene ID: 27421. Target Synonyms: ABC-C6; ABC34; ARA; MLP1; MOATE; MRP6; PXE; PXE1; Pseudoxanthoma Elasticum. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5879