Learn More
Biomatik Corporation Mouse ATPase, Na+/K+ Transporting Beta 1 Polypeptide (ATP1b1) ELISA Kit
Supplier:  Biomatik Corporation EKU02634
Encompass_Preferred
Mouse ATPase, Na+/K+ Transporting Beta 1 Polypeptide (ATP1b1) ELISA Kit for detecting ATP1b1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q545P0. Gene ID: 11931. Target Synonyms: ATP1B; Sodium/potassium-dependent ATPase subunit beta-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9109