Learn More
Biomatik Corporation Mouse Atrial Natriuretic Peptide (ANP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11613
Encompass_Preferred
Mouse Atrial Natriuretic Peptide (ANP) Mini sample ELISA Kit for detecting ANP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.64pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05125. Gene ID: 230899. Target Synonyms: ANH; ANF; CDD; Atrial Natriuretic Factor; Atrial Natriuretic Hormone; Atriopeptin; Cardionatrine; Cardiodilatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8119